Gene Transcripts

Gene Symbol
Rltpr
Entrez Gene ID
307797
Gene Name
RGD motif, leucine rich repeats, tropomodulin domain and proline-rich containing
Gene Aliases
RGD1562390
Chromosome Location
Chr. 19: 37652384 - 37665333 on Build Rnor_6.0
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq XM_006255548.2 XP_006255610.1 9 1342
XM_008772559.1 XP_008770781.1 9 1342

View Full Product Details