Gene Transcripts

Gene Symbol
Arid5b
Entrez Gene ID
309728
Gene Name
AT rich interactive domain 5B (Mrf1 like)
Gene Aliases
Chromosome Location
Chr. 20: 21713900 - 21881193 on Build Rnor_6.0
UniGene ID
Rn.204911
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001107624.1 NP_001101094.1 2011
GenBank mRNA FQ229049.1 - 5 1114

View Full Product Details