Gene Transcripts

Gene Symbol
Aadacl4
Entrez Gene ID
313688
Gene Name
arylacetamide deacetylase-like 4
Gene Aliases
RGD1565761
Chromosome Location
Chr. 5: 162676360 - 162688866 on Build Rnor_6.0
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq XM_233640.4 XP_233640.4 2, 3 378

View Full Product Details