Gene Transcripts

Gene Symbol
Shroom4
Entrez Gene ID
317391
Gene Name
shroom family member 4
Gene Aliases
RGD1563434
Chromosome Location
Chr. X: 16719818 - 16929829 on Build Rnor_6.0
UniGene ID
Rn.136522
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001191730.1 NP_001178659.1 9 4286
XM_006256763.2 XP_006256825.1 8 4363

View Full Product Details