Gene Transcripts

Gene Symbol
Gsx1
Entrez Gene ID
288457
Gene Name
GS homeobox 1
Gene Aliases
Gsh1
Chromosome Location
Chr. 12: 9605873 - 9607168 on Build Rnor_6.0
UniGene ID
Rn.228201
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001191663.1 NP_001178592.1 2 705

View Full Product Details