Gene Transcripts

Gene Symbol
Csn1s2a
Entrez Gene ID
114595
Gene Name
casein alpha s2-like A
Gene Aliases
Csng
Chromosome Location
Chr. 14: 21930858 - 21944921 on Build Rnor_6.0
UniGene ID
Rn.92369
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001105741.1 NP_001099211.1 12 557
GenBank mRNA J00712.1 - 12 557

View Full Product Details