Gene Transcripts

Gene Symbol
Ttc36
Entrez Gene ID
300676
Gene Name
tetratricopeptide repeat domain 36
Gene Aliases
Chromosome Location
Chr. 8: 49106374 - 49109981 on Build Rnor_6.0
UniGene ID
Rn.107180
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001005546.1 NP_001005546.1 2 374
GenBank mRNA BC082024.1 - 2 374
FQ210191.1 - 2 322
FQ211414.1 - 2 322

View Full Product Details