Gene Transcripts

Gene Symbol
Olr82
Entrez Gene ID
365328
Gene Name
olfactory receptor 82
Gene Aliases
Chromosome Location
Chr. 1: 168309127 - 168310906 on Build Rnor_6.0
UniGene ID
Rn.141904
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001001282.1 NP_001001282.1 1 912
XM_006229914.1 XP_006229976.1 2 945

View Full Product Details