Gene Transcripts

Gene Symbol
Vom2r22
Entrez Gene ID
690758
Gene Name
vomeronasal 2 receptor, 22
Gene Aliases
Chromosome Location
Chr. 8: 4349354 - 4376039 on Build Rnor_6.0
UniGene ID
Rn.198478
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001099656.1 NP_001093126.1 2532
XM_003750425.1 XP_003750473.1 6 2532

View Full Product Details