Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
NMBR
Entrez Gene ID
574262
Gene Name
neuromedin B receptor
Gene Aliases
EGK_21259
Chromosome Location
Chr.4: 45822517 - 45835811 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001278400.1 NP_001265329.1 - 435 81
GenBank mRNA AY350448.1 - - 239 81

View Full Product Details