Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ACSF2
Entrez Gene ID
702474
Gene Name
acyl-CoA synthetase family member 2
Gene Aliases
-
Chromosome Location
Chr.16: 46312490 - 46361213 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_001097818.4 XP_001097818.2 - 1420 74
XM_015119306.2 XP_014974792.2 - 1258 74
GenBank mRNA JV072084.1 - - 1241 74
JV662150.1 - - 1244 74

View Full Product Details