Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
TUBE1
Entrez Gene ID
696810
Gene Name
tubulin epsilon 1
Gene Aliases
-
Chromosome Location
Chr.4: 15706146 - 15723073 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_015137557.2 XP_014993043.1 - 1618 97
XM_015137561.2 XP_014993047.1 - 3182 97
XM_028847048.1 XP_028702881.1 - 1389 97
XM_028847049.1 XP_028702882.1 - 1356 97
XM_028847050.1 XP_028702883.1 - 1353 97
GenBank mRNA JV637581.1 - - 1338 97

View Full Product Details