Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
TRAPPC1
Entrez Gene ID
716622
Gene Name
trafficking protein particle complex 1
Gene Aliases
EGK_08154
Chromosome Location
Chr.16: 7724459 - 7726092 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001194511.2 NP_001181440.1 - 272 65
XM_001111285.4 XP_001111285.1 - 230 65
GenBank mRNA JU320131.1 - - 234 65
JU320132.1 - - 233 65
JU473417.1 - - 263 65
JV691979.1 - - 256 65

View Full Product Details