Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
SRPX
Entrez Gene ID
697145
Gene Name
sushi repeat containing protein X-linked
Gene Aliases
EGK_20388
Chromosome Location
Chr.X: 37963001 - 38037172 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_001083979.4 XP_001083979.2 - 888 85
XM_001084091.4 XP_001084091.2 - 1092 85
GenBank mRNA JU327493.1 - - 620 85
JV636407.1 - - 877 85

View Full Product Details