Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
SNTG2
Entrez Gene ID
706376
Gene Name
syntrophin gamma 2
Gene Aliases
-
Chromosome Location
Chr.13: 107398833 - 107768591 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_028832025.1 XP_028687858.1 - 994 73
XM_028832026.1 XP_028687859.1 - 1011 73

View Full Product Details