Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
PPP6R3
Entrez Gene ID
710120
Gene Name
protein phosphatase 6 regulatory subunit 3
Gene Aliases
EGK_05908, SAPS3
Chromosome Location
Chr.14: 6149374 - 6307267 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_015113490.2 XP_014968976.1 - 2063 142
XM_015113492.2 XP_014968978.1 - 2045 142
XM_015113495.2 XP_014968981.1 - 1976 142
XM_028833114.1 XP_028688947.1 - 1770 142
XM_028833115.1 XP_028688948.1 - 1737 142
XM_028833116.1 XP_028688949.1 - 1788 142
XM_028833117.1 XP_028688950.1 - 1770 142
XM_028833118.1 XP_028688951.1 - 1701 142
XM_028833119.1 XP_028688952.1 - 1683 142
XM_028833120.1 XP_028688953.1 - 1737 142
XM_028833121.1 XP_028688954.1 - 1737 142
XM_028833122.1 XP_028688955.1 - 1650 142
XM_028833123.1 XP_028688956.1 - 1635 142
XM_028833124.1 XP_028688957.1 - 1701 142
XM_028833125.1 XP_028688958.1 - 1788 142
XM_028833126.1 XP_028688959.1 - 1683 142
XM_028833127.1 XP_028688960.1 - 1683 142
XM_028833128.1 XP_028688961.1 - 2063 142
XM_028833129.1 XP_028688962.1 - 2412 142
XM_028833130.1 XP_028688963.1 - 2161 142
XM_028833131.1 XP_028688964.1 - 2128 142
XM_028833132.1 XP_028688965.1 - 2160 142
XM_028833133.1 XP_028688966.1 - 1989 142
XM_028833134.1 XP_028688967.1 - 1650 142
XM_028833136.1 XP_028688969.1 - 1584 142
XM_028833137.1 XP_028688970.1 - 1548 142
GenBank mRNA JU321666.1 - - 1848 142
JU321700.1 - - 1935 142
JU321705.1 - - 1935 142
JU321706.1 - - 1935 142
JU321718.1 - - 1935 142
JU471603.1 - - 2045 142
JU471604.1 - - 1958 142
JV047549.1 - - 1996 142
JV047550.1 - - 1909 142

View Full Product Details