Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
EMSY
Entrez Gene ID
697319
Gene Name
EMSY transcriptional repressor, BRCA2 interacting
Gene Aliases
C14H11orf30, EGK_06698
Chromosome Location
Chr.14: 69507060 - 69611882 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_015115228.2 XP_014970714.1 - 2377 110
XM_015115229.2 XP_014970715.1 - 2374 110
XM_015115230.2 XP_014970716.1 - 2350 110
XM_015115231.2 XP_014970717.1 - 2377 110
XM_015115232.2 XP_014970718.1 - 2347 110
XM_015115233.2 XP_014970719.1 - 2224 110
XM_015115236.2 XP_014970722.1 - 2197 110
XM_015115237.2 XP_014970723.1 - 2224 110
XM_015115239.2 XP_014970725.1 - 2350 110
XM_028833834.1 XP_028689667.1 - 2377 110
GenBank mRNA JU335540.1 - - 2328 110
JU476467.1 - - 2328 110
JV719156.1 - - 2333 110

View Full Product Details