Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
PCK1
Entrez Gene ID
696207
Gene Name
phosphoenolpyruvate carboxykinase 1
Gene Aliases
EGK_02261
Chromosome Location
Chr.10: 92734769 - 92740427 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_001086710.4 XP_001086710.2 - 563 76

View Full Product Details