Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
LOC711810
Entrez Gene ID
711810
Gene Name
lithostathine-1-alpha
Gene Aliases
EGK_05495
Chromosome Location
Chr.13: 29508840 - 29511781 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_015112727.2 XP_014968213.1 - 134 131

View Full Product Details