Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ARFRP1
Entrez Gene ID
695611
Gene Name
ADP ribosylation factor related protein 1
Gene Aliases
EGK_02179
Chromosome Location
Chr.10: 98909089 - 98917671 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_001084614.4 XP_001084614.2 - 418 87
XM_015148593.2 XP_015004079.1 - 413 87
XM_015148595.2 XP_015004081.1 - 345 87
GenBank mRNA JU332425.1 - - 266 87
JU332426.1 - - 266 87
JU473884.1 - - 266 87
JU473885.1 - - 266 87
JV044112.1 - - 266 87
JV044113.1 - - 266 87
JV603000.1 - - 400 87
JV704050.1 - - 266 87

View Full Product Details