Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
SACS
Entrez Gene ID
706831
Gene Name
sacsin molecular chaperone
Gene Aliases
sacsin
Chromosome Location
Chr.17: 3266959 - 3374442 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_015120738.2 XP_014976224.2 - 445 95
XM_028837425.1 XP_028693258.1 - 971 95
XM_028837426.1 XP_028693259.1 - 971 95

View Full Product Details