Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
JADE2
Entrez Gene ID
713338
Gene Name
jade family PHD finger 2
Gene Aliases
EGK_16861, PHF15
Chromosome Location
Chr.6: 131986624 - 132045049 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_015140931.2 XP_014996417.1 - 537 134
XM_015140932.2 XP_014996418.1 - 538 134
XM_015140933.2 XP_014996419.1 - 540 134
XM_015140934.2 XP_014996420.1 - 541 134
XM_015140935.2 XP_014996421.1 - 669 134
XM_015140936.2 XP_014996422.1 - 561 134
XM_015140937.2 XP_014996423.1 - 394 134
XM_015140939.2 XP_014996425.1 - 902 134
XM_015140940.2 XP_014996426.1 - 430 134
XM_015140941.2 XP_014996427.1 - 669 134
GenBank mRNA JU333651.1 - - 407 134
JU474902.1 - - 326 134

View Full Product Details