Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
RMDN1
Entrez Gene ID
694237
Gene Name
regulator of microtubule dynamics 1
Gene Aliases
FAM82B
Chromosome Location
Chr.8: 86893999 - 86929626 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_001083289.4 XP_001083289.1 - 879 136
XM_015145669.2 XP_015001155.1 - 876 136

View Full Product Details