Assay Design
The assay probe spans an exon junction.
Gene Transcripts
- Gene Symbol
- HYI
- Entrez Gene ID
-
700926
- Gene Name
- hydroxypyruvate isomerase (putative)
- Gene Aliases
-
-
- Chromosome Location
-
Chr.1: 181012465 - 181015523 on Build Mmul_10
- Gene Symbol
- SZT2
- Entrez Gene ID
-
702661
- Gene Name
- SZT2 subunit of KICSTOR complex
- Gene Aliases
-
-
- Chromosome Location
-
Chr.1: 181015522 - 181077986 on Build Mmul_10
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
GenBank mRNA |
JV576352.1 |
- |
- |
7463 |
105 |
View Full Product Details