Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
UCK1
Entrez Gene ID
715417
Gene Name
uridine-cytidine kinase 1
Gene Aliases
-
Chromosome Location
Chr.15: 8892079 - 8899959 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001194319.3 NP_001181248.1 - 591 97
GenBank mRNA JU332166.1 - - 576 97
JU473602.1 - - 523 97
JV044854.1 - - 611 97

View Full Product Details