Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
C19H19orf47
Entrez Gene ID
701191
Gene Name
chromosome 19 C19orf47 homolog
Gene Aliases
-
Chromosome Location
Chr.19: 39526156 - 39555142 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
GenBank mRNA JU332154.1 - - 734 104
JU473803.1 - - 571 104
JV045718.1 - - 617 104

View Full Product Details