Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
METTL7A
Entrez Gene ID
693894
Gene Name
methyltransferase like 7A
Gene Aliases
EGK_03645
Chromosome Location
Chr.11: 50547130 - 50556503 on Build Mmul_10
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001193711.2 NP_001180640.2 - 76 73

View Full Product Details