Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Nmbr
Entrez Gene ID
25264
Gene Name
neuromedin B receptor
Gene Aliases
NMB-R
Chromosome Location
Chr.1: 8933770 - 8945240 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_012799.1 NP_036931.1 1 - 2 551 115
GenBank mRNA U37058.1 - 1 - 2 551 115

View Full Product Details