The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_012878.2 | NP_037010.1 | 1 - 2 | 212 | 118 |
XM_008771065.1 | XP_008769287.1 | 1 - 2 | 215 | 118 | |
GenBank mRNA | BC070507.1 | - | 2 - 3 | 221 | 118 |
M81231.1 | - | 2 - 3 | 219 | 118 |