Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Tub
Entrez Gene ID
25609
Gene Name
tubby bipartite transcription factor
Gene Aliases
-
Chromosome Location
Chr.1: 173550537 - 173632194 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_013077.1 NP_037209.1 3 - 4 250 136
XM_008759690.1 XP_008757912.1 3 - 4 526 136
XM_008759691.1 XP_008757913.1 3 - 4 527 136
GenBank mRNA AB011544.1 - 3 - 4 250 136

View Full Product Details