The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001160162.1 | NP_001153634.1 | 21 - 22 | 4005 | 54 |
NM_013125.2 | NP_037257.1 | 22 - 23 | 4164 | 54 | |
XM_006244076.2 | XP_006244138.1 | 22 - 23 | 4334 | 54 | |
XM_008766670.1 | XP_008764892.1 | 22 - 23 | 5362 | 54 | |
XM_008766671.1 | XP_008764893.1 | 21 - 22 | 5203 | 54 | |
GenBank mRNA | AF353637.1 | - | 20 - 21 | 3808 | 54 |
M27902.1 | - | 22 - 23 | 4164 | 54 |