The assay primers and probes lie within a single exon. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_021700.1 | NP_067732.1 | 1 - 1 | 1048 | 70 |
GenBank mRNA | Y10619.1 | - | 1 - 1 | 1048 | 70 |