The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_021848.1 | NP_068620.1 | 11 - 12 | 1287 | 78 |
GenBank mRNA | AF105368.1 | - | 11 - 12 | 1287 | 78 |