The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_030845.1 | NP_110472.1 | 3 - 4 | 323 | 102 |
GenBank mRNA | D11444.1 | - | 3 - 4 | 308 | 102 |
M86536.1 | - | 3 - 4 | 323 | 102 |