Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Cxxc4
Entrez Gene ID
83824
Gene Name
CXXC finger protein 4
Gene Aliases
Idax
Chromosome Location
Chr.2: 239414848 - 239439640 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_008761530.1 XP_008759752.1 2 - 3 1504 115
GenBank mRNA AF272158.1 - 1 - 2 567 115

View Full Product Details