The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_130819.1 | NP_570832.1 | 3 - 4 | 746 | 65 |
GenBank mRNA | AF337953.1 | - | 3 - 4 | 746 | 65 |