Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Ctbp2
Entrez Gene ID
81717
Gene Name
C-terminal binding protein 2
Gene Aliases
-
Chromosome Location
Chr.1: 205001354 - 205030565 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_053335.1 NP_445787.1 4 - 5 2257 61
GenBank mRNA AF222712.1 - 4 - 5 2257 61

View Full Product Details