The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_178096.3 | NP_835197.1 | 2 - 3 | 134 | 84 |
XM_006254595.1 | XP_006254657.1 | 3 - 4 | 233 | 84 | |
GenBank mRNA | AB072344.1 | - | 2 - 3 | 139 | 84 |
AY724475.1 | - | 3 - 4 | 165 | 84 | |
FQ214488.1 | - | 3 - 4 | 294 | 84 |