The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_181480.1 | NP_852145.1 | 5 - 6 | 409 | 69 |
XM_008760144.1 | XP_008758366.1 | 4 - 5 | 436 | 69 | |
XM_008760145.1 | XP_008758367.1 | 3 - 4 | 182 | 69 | |
GenBank mRNA | AB090805.1 | - | 5 - 6 | 409 | 69 |
BC161832.1 | - | 5 - 6 | 483 | 69 |