Assay Design
The assay probe spans an exon junction. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- Vegp2
- Entrez Gene ID
-
94106
- Gene Name
- von Ebners gland protein 2
- Gene Aliases
-
-
- Chromosome Location
-
Chr.3: 4217260 - 4221330 on Build Rnor_6.0
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_053574.1 |
NP_446026.1 |
6 - 7 |
588 |
64 |
GenBank mRNA |
X74806.1 |
- |
6 - 7 |
588 |
64 |
- Gene Symbol
- LOC100911507
- Entrez Gene ID
-
100911507
- Gene Name
- von Ebner gland protein 2-like
- Gene Aliases
-
-
- Chromosome Location
-
Chr.3: 4137456 - 4140236 on Build Rnor_6.0
View Full Product Details