The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_013053.1 | NP_037185.1 | 4 - 5 | 660 | 103 |
GenBank mRNA | BC062409.1 | - | 3 - 4 | 630 | 103 |
D17614.1 | - | 4 - 5 | 660 | 103 | |
FQ218679.1 | - | 3 - 4 | 420 | 103 |