The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_031353.1 | NP_112643.1 | 8 - 9 | 824 | 112 |
XM_006246346.1 | XP_006246408.1 | 7 - 8 | 797 | 112 | |
GenBank mRNA | AB039662.1 | - | 8 - 9 | 824 | 112 |
AF048828.1 | - | 8 - 9 | 868 | 112 | |
AF268467.1 | - | 7 - 8 | 758 | 112 | |
BC060558.1 | - | 8 - 9 | 801 | 112 | |
BC072484.1 | - | 8 - 9 | 808 | 112 | |
BC087573.1 | - | 8 - 9 | 803 | 112 | |
BC087657.1 | - | 8 - 9 | 780 | 112 | |
BC104684.1 | - | 8 - 9 | 794 | 112 | |
BC127491.1 | - | 8 - 9 | 780 | 112 |