The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001039325.1 | NP_001034414.1 | 2 - 3 | 707 | 72 |
GenBank mRNA | AY886537.1 | - | 2 - 3 | 707 | 72 |