The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_013200.1 | NP_037332.1 | 18 - 19 | 2277 | 71 |
GenBank mRNA | BC085761.1 | - | 18 - 19 | 2289 | 71 |
D43623.1 | - | 17 - 18 | 2264 | 71 |