Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Slc22a16
Entrez Gene ID
682934
Gene Name
solute carrier family 22 (organic cation/carnitine transporter), member 16
Gene Aliases
-
Chromosome Location
Chr.20: 45658398 - 45669192 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_003751977.3 XP_003752025.2 1 - 2 157 96

View Full Product Details