Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Klhl33
Entrez Gene ID
100362676
Gene Name
kelch-like family member 33
Gene Aliases
-
Chromosome Location
Chr.15: 27811226 - 27819806 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_002728195.4 XP_002728241.3 4 - 4 4903 138

View Full Product Details