Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Susd4
Entrez Gene ID
289335
Gene Name
sushi domain containing 4
Gene Aliases
RGD1564043
Chromosome Location
Chr.13: 101181664 - 101307912 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001105982.1 NP_001099452.1 2 - 3 319 63
XM_006250373.2 XP_006250435.1 2 - 3 473 63
XM_006250375.2 XP_006250437.1 2 - 3 358 63
XM_008769820.1 XP_008768042.1 2 - 3 473 63
XM_008769821.1 XP_008768043.1 2 - 3 362 63
XM_008769822.1 XP_008768044.1 2 - 3 451 63
XM_008769823.1 XP_008768045.1 2 - 3 473 63
GenBank mRNA FQ212123.1 - 2 - 3 303 63

View Full Product Details