The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001107964.1 | NP_001101434.1 | 2 - 3 | 266 | 110 |
XM_006238665.2 | XP_006238727.1 | 2 - 3 | 856 | 110 | |
GenBank mRNA | FQ219156.1 | - | 2 - 3 | 220 | 110 |