Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Hyi
Entrez Gene ID
500536
Gene Name
hydroxypyruvate isomerase
Gene Aliases
RGD1561416
Chromosome Location
Chr.5: 137189443 - 137192124 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_002729532.4 XP_002729578.2 1 - 2 238 90
XM_006238746.2 XP_006238808.1 1 - 2 240 90
XM_006238747.2 XP_006238809.1 1 - 2 240 90
XM_006238748.2 XP_006238810.1 1 - 2 240 90
GenBank mRNA FQ211344.1 - 1 - 2 221 90

View Full Product Details