The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001080148.1 | NP_001073617.1 | 1 - 2 | 230 | 51 |
GenBank mRNA | AY310319.1 | - | 1 - 2 | 230 | 51 |
AY921220.2 | - | 1 - 2 | 292 | 51 |